Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylfluorene 98.0+%, TCI America™
SDP

Catalog No. A10695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10695G 5 g
A106925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10695G Supplier TCI America Supplier No. A10695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 781-73-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00001124
InChI Key IBASEVZORZFIIH-UHFFFAOYSA-N
Synonym 2-acetylfluorene, 1-9h-fluoren-2-yl ethanone, ethanone, 1-9h-fluoren-2-yl, 2-fluorenyl methyl ketone, 2-acetofluorene, ketone, fluoren-2-yl methyl, fluoren-2-yl methyl ketone, 1-9h-fluoren-2-yl ethan-1-one, 2-acetyl-9h-fluorene
PubChem CID 69908
IUPAC Name 1-(9H-fluoren-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C3=CC=CC=C3C2

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylfluorene
RTECSNumber OB3800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.