Learn More
Medchemexpress LLC 2-acetylnaphthalene | 93-08-3 | MFCD00004108 | 25g
SDP
Catalog No. 5000266334
Encompass_Preferred
2-Acetonaphthone is a synthetic fragrance material 2-Acetonaphthone increases ROS under UVA/sunlight leading to endoplasmic reticulum stress and decreased mitochondrial membrane potential 2-Acetonaphthone can be used as an adulterant in a variety of cosmetics 2-Acetonaphthone can be used for the study of skin keratinization[1]
Catalog No. 50-002-66334 Supplier Medchemexpress LLC Supplier No. HYY181925G
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2-Acetonaphthone is a synthetic fragrance material 2-Acetonaphthone increases ROS under UVA/sunlight leading to endoplasmic reticulum stress and decreased mitochondrial membrane potential 2-Acetonaphthone can be used as an adulterant in a variety of cosmetics 2-Acetonaphthone can be used for the study of skin keratinization[1]