Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Acetylpyridine

Catalog No. A21002100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21002100G 100 g
A2100225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21002100G Supplier Sigma Aldrich Supplier No. A21002100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-acetylpyridine can be used in the synthesis of:
  • A near-infrared fluorescent probe for monitoring pH changes in living cells
  • Biologically active ligands for preparing various metal complexes including co(ii), ni(ii), cu(ii) and zn(ii)
  • A water-soluble, time-gated luminescence probe for singlet oxygen detection
  • 2-acetylpyridine thiosemicarbazones are potent iron chelators and antiproliferative agents

Specifications

CAS 1122-62-9
Density 1.08 g/mL (at 25°C (literature))
Boiling Point 188°C to 189°C (lit.)
Molecular Formula C7H7NO
Refractive Index n20/D 1.521 (literature)
Linear Formula C7H7NO
MDL Number MFCD00006303
Quantity 100 g
Synonym Methyl 2-pyridyl ketone
Molecular Weight (g/mol) 121.14
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.