Learn More
Medchemexpress LLC 2-Acetylpyrrole | 1072-83-9 | 100.0% | 109.13 | 1 ML
SDP
Catalog No. 5000367000
Encompass_Preferred
2-Acetylpyrrole is an organic compound featuring a pyrrole ring. It is generated during the Maillard browning reaction and acts as a key flavor component in various foods. Additionally, this compound functions as an intermediate in the creation of other active compounds, including the flavor compound 2-acetyl-1-pyrroline.
  • It is an organic compound containing a pyrrole ring.
  • It is a product of the Maillard browning reaction.
  • It serves as a major flavor component in many foods.
  • It can be used as an intermediate in the synthesis of other active compounds.
Catalog No. 50-003-67000 Supplier Medchemexpress LLC Supplier No. HYW01295610MM/1ML
2-Acetylpyrrole is an organic compound featuring a pyrrole ring. It is generated during the Maillard browning reaction and acts as a key flavor component in various foods. Additionally, this compound functions as an intermediate in the creation of other active compounds, including the flavor compound 2-acetyl-1-pyrroline.
  • It is an organic compound containing a pyrrole ring.
  • It is a product of the Maillard browning reaction.
  • It serves as a major flavor component in many foods.
  • It can be used as an intermediate in the synthesis of other active compounds.