Learn More
Medchemexpress LLC 2-Acetylpyrrole | 1072-83-9 | 99.98% | 109.13 | 25 G
SDP
Catalog No. 5000453835
Encompass_Preferred
2-Acetylpyrrole is an organic compound containing a pyrrole ring. It is a product of the Maillard browning reaction and acts as a major flavor component in many foods. It can also be used as an intermediate in the synthesis of other active compounds, such as 2-acetyl-1-pyrroline. This product is for research use only.
  • Organic compound with a pyrrole ring
  • Product of Maillard browning reaction
  • Major flavor component in many foods
  • Intermediate in synthesis of other active compounds
  • Solid, white to off-white appearance
Catalog No. 50-004-53835 Supplier Medchemexpress LLC Supplier No. 25GHYW012956
2-Acetylpyrrole is an organic compound containing a pyrrole ring. It is a product of the Maillard browning reaction and acts as a major flavor component in many foods. It can also be used as an intermediate in the synthesis of other active compounds, such as 2-acetyl-1-pyrroline. This product is for research use only.
  • Organic compound with a pyrrole ring
  • Product of Maillard browning reaction
  • Major flavor component in many foods
  • Intermediate in synthesis of other active compounds
  • Solid, white to off-white appearance