Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt solution

Catalog No. 655821250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-582-1250ML 250 mL
65-582-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-582-1250ML Supplier Sigma Aldrich Supplier No. 655821250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt (AMPSNa) is a hydrophilic compound that can be used as a dopant and a protonating agent for conducting polymers. It is used in a variety of electronic applications.AMPSNa can be used in the formation of polyelectrolyte copolymer gels for potential application in bioengineering, biomedicine and water purification. It can also be used in the fabrication of Schottky diodes, humidity sensors, and lithium ion batteries.

Specifications

CAS 5165-97-9
Density 1.2055 g/mL (at 25°C)
Molecular Formula C7H12NNaO4S
Refractive Index n20/D 1.4220 (literature)
Linear Formula H2C=CHCONHC(CH3)2 CH2SO3Na
Quantity 250 mL
MDL Number MFCD00156481
Synonym 2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid sodium; Sodium 2-acrylamido-2-methyl-1-propanesulfonate; Sodium acryloyldimethyltaurate
Molecular Weight (g/mol) 229.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.