Learn More
Embi Tec 2% Agarose TAE w/ GelGreen Long Gel 4(24+1) Well
SDP
Catalog No. NC1849788
Encompass
Just open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.
Catalog No. NC1849788 Supplier Embi Tec Supplier No. EP1118
Just open gel package and place gel with tray directly onto the RunOne Unit platform or similar size platform.