Learn More
Embi Tec 2% AGAROSE TBE GEL 8(12+1)WELL
SDP
Catalog No. NC0136411
Encompass
2% Agarose TBE w/EtBr Long Gel, 8(12+1) well, 5/box
Catalog No. NC0136411 Supplier Embi Tec Supplier No. GE4672
2% Agarose TBE w/EtBr Long Gel, 8(12+1) well, 5/box