Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Allyl 4-ethyl 5-amino-3-methylthiophene-2,4-dicarboxylate

Catalog No. OTV000545 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000545 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000545 Supplier Sigma Aldrich Supplier No. OTV000545250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Comparision of electrochemical polymerization properties of 3-thiophenemethanol and 3-methylthiophene has been reported.3-thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays
  • It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile

Specifications

Molecular Formula C12H15NO4S
Linear Formula C12H15NO4S
Quantity 5 g
MDL Number MFCD04065873
Molecular Weight (g/mol) 269.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.