Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allyl-D-glycine, 95%

Catalog No. AAH52112MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52112MD 250 mg
AAH5211203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52112MD Supplier Thermo Scientific Chemicals Supplier No. H52112MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 54594-06-8 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54594-06-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00063104
InChI Key WNNNWFKQCKFSDK-SCSAIBSYSA-N
Synonym d-allylglycine, r-2-aminopent-4-enoic acid, r-2-amino-4-pentenoic acid, 2r-2-aminopent-4-enoic acid, h-d-gly allyl-oh, r-2-amino-pent-4-enoic acid, 4-pentenoic acid, 2-amino-, 2r, 2r-2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, r-allylglycine
PubChem CID 6992334
IUPAC Name (2R)-2-aminopent-4-enoic acid
SMILES C=CCC(C(=O)O)N

Specifications

Quantity 250 mg
Formula Weight 115.13
Percent Purity 95%
Chemical Name or Material 2-Allyl-D-glycine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.