Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allyloxytetrahydropyran 95.0+%, TCI America™
SDP

Catalog No. A23915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23915G 5 g
A239125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23915G Supplier TCI America Supplier No. A23915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4203-49-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00154089
InChI Key NRAKYXXDEXCVMV-UHFFFAOYNA-N
Synonym 2-allyloxytetrahydropyran, 2-allyloxy tetrahydro-2h-pyran, 2-allyloxy tetrahydropyran, 2-prop-2-en-1-yloxy oxane, acmc-209jmz, 2-allyloxy-tetrahydro-pyran, allyl tetrahydropyranyl ether, ksc490q3f, tetrahydro-2-allyloxy-2h-pyran
PubChem CID 4532059
IUPAC Name 2-(prop-2-en-1-yloxy)oxane
SMILES C=CCOC1CCCCO1

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 g
UN Number 3271
Formula Weight 142.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Allyloxytetrahydropyran
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.