Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allylphenol, 98+%

Catalog No. aaa1075522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1075522 100 g
AAA1075536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1075522 Supplier Thermo Scientific Chemicals Supplier No. A1075522
Only null left
Add to Cart

CAS: 1745-81-9 | C9H10O | 134.178 g/mol

2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

Solubility
Soluble in water(7 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1745-81-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002250
InChI Key QIRNGVVZBINFMX-UHFFFAOYSA-N
Synonym 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
PubChem CID 15624
ChEBI CHEBI:39826
IUPAC Name 2-prop-2-enylphenol
SMILES C=CCC1=CC=CC=C1O

Specifications

Melting Point -6°C
Density 1.024
Boiling Point 220°C to 222°C
Flash Point 88°C (190°F)
Odor Phenol-like
Refractive Index 1.545
Quantity 100 g
UN Number UN2922
Beilstein 742121
Solubility Information Soluble in water(7g/L).
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 2-Allylphenol
Show More Show Less
Hazard Category H227-H302+H312-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 217-119-0
RTECSNumber SJ3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.