Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Allylphenoxy)trimethylsilane, 98%, Thermo Scientific™

Catalog No. AAH3090522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3090522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3090522 Supplier Thermo Scientific Chemicals Supplier No. H3090522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18042-43-8
Molecular Formula C12H18OSi
Molecular Weight (g/mol) 206.36
MDL Number MFCD12407078
InChI Key LJIXCUHZKDSCQX-UHFFFAOYSA-N
Synonym 2-allylphenoxy trimethylsilane, trimethyl 2-prop-2-en-1-yl phenoxy silane, o-allylphenoxytrimethylsilane, trimethyl-2-prop-2-enylphenoxy silane, silane, trimethyl 2-2-propenyl phenoxy
PubChem CID 15049460
IUPAC Name trimethyl-(2-prop-2-enylphenoxy)silane
SMILES C[Si](C)(C)OC1=CC=CC=C1CC=C

Specifications

Boiling Point 85°C (5mmHg)
Quantity 100 g
Formula Weight 206.36
Percent Purity 98%
Sensitivity Moisture sensitive
Density 0.925
Chemical Name or Material (2-Allylphenoxy)trimethylsilane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.