Learn More
Medchemexpress LLC 2-Amino-1,3-bis(carboxylethoxy)propane hydrochloride | 2421153-68-4 | ≥97.0% | 271.70 | 100 MG
SDP
Catalog No. 5000400858
Encompass_Preferred
2-Amino-1,3-bis(carboxylethoxy)propane hydrochloride is a non-protein amino acid, sulfamic acid. It contains one amino group and two terminal carboxylic acids. This compound has a variety of physiological functions and is for research use only.
  • Non-protein amino acid
  • Sulfamic acid
  • Contains one amino group and two terminal carboxylic acids
  • Maintains stability of cell membranes
  • Regulates cholesterol metabolism
  • Supports normal function of the nervous system
  • Participates in collagen synthesis
  • Anti-oxidation
Catalog No. 50-004-00858 Supplier Medchemexpress LLC Supplier No. HY23212A100MG
2-Amino-1,3-bis(carboxylethoxy)propane hydrochloride is a non-protein amino acid, sulfamic acid. It contains one amino group and two terminal carboxylic acids. This compound has a variety of physiological functions and is for research use only.
  • Non-protein amino acid
  • Sulfamic acid
  • Contains one amino group and two terminal carboxylic acids
  • Maintains stability of cell membranes
  • Regulates cholesterol metabolism
  • Supports normal function of the nervous system
  • Participates in collagen synthesis
  • Anti-oxidation