Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1,3-propanediol, 98%

Catalog No. p-3756847
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401030050 5 g Glass bottle
AC401030250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401030050 Supplier Thermo Scientific Chemicals Supplier No. 401030050
Only null left
Add to Cart

CAS: 534-03-2 | C3H9NO2 | 91.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-03-2
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00051487
InChI Key KJJPLEZQSCZCKE-UHFFFAOYSA-N
Synonym 2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
PubChem CID 68294
IUPAC Name 2-aminopropane-1,3-diol
SMILES NC(CO)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 52.0°C to 56.0°C
Color White
Boiling Point 115.0°C to 116.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2CH(CH2OH)2
Quantity 5 g
Formula Weight 91.11
Percent Purity 98%
Physical Form Adhering Crystalline Powder
Chemical Name or Material 2-Amino-1, 3-propanediol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-584-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.