Learn More
Accela Chembio Inc 2-amino-1-morpholinoethanone | 1g | 56414-96-1 | MFCD06658935 | 95+% | Shelf Life: 900 Days | Light Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY0103451G
Encompass_Preferred
2-amino-1-morpholinoethanone | 1g | 56414-96-1 | MFCD06658935 | 95+% | Shelf Life: 900 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-354-907