Learn More
Apexbio Technology LLC 2-Amino-1-phenylethanol 7568-93-6 1g
SDP
Catalog No. 5000092444
Encompass_Preferred
2-Amino-1-phenylethanol (CAS 7568-93-6) is an organic compound structurally related to norepinephrine featuring both amino and hydroxyl functional groups Due to these characteristics it serves as a useful analogue for investigating catecholaminergic neurotransmission In vitro studies utilize this molecule to examine ligand-receptor interactions among adrenergic receptor subtypes providing insights into the signal transduction pathways associated with catecholamine activity It is widely applied in fundamental research within neuropharmacology and drug development targeting adrenergic mechanisms
Catalog No. 50-000-92444 Supplier Apexbio Technology LLC Supplier No. C65841000
2-Amino-1-phenylethanol (CAS 7568-93-6) is an organic compound structurally related to norepinephrine featuring both amino and hydroxyl functional groups Due to these characteristics it serves as a useful analogue for investigating catecholaminergic neurotransmission In vitro studies utilize this molecule to examine ligand-receptor interactions among adrenergic receptor subtypes providing insights into the signal transduction pathways associated with catecholamine activity It is widely applied in fundamental research within neuropharmacology and drug development targeting adrenergic mechanisms