Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2-(2-fluorophenyl)acetic acid | 84145-28-8 | MFCD00042726 | 99.9% | 169.16 g/mol | C8H8FNO2 | 25 G
SDP

Catalog No. 5000302214
Encompass_Preferred

2-Amino-2-(2-fluorophenyl)acetic acid is an amino-acid derivative used in research applications as a glycine analogue. The material is supplied as a solid with analytical documentation and recommended storage conditions to support experimental workflows.

  • High purity: 99.92%, suitable for research use.
  • Includes certificate of analysis (COA), safety data sheet (SDS), H NMR, and MS files.
  • Appearance: off-white to light-brown solid, easy to weigh and handle.
  • Storage: store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only.

Catalog No. 50-003-02214 Supplier Medchemexpress LLC Supplier No. 25GHYW026894
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2-(2-fluorophenyl)acetic acid is an amino-acid derivative used in research applications as a glycine analogue. The material is supplied as a solid with analytical documentation and recommended storage conditions to support experimental workflows.

  • High purity: 99.92%, suitable for research use.
  • Includes certificate of analysis (COA), safety data sheet (SDS), H NMR, and MS files.
  • Appearance: off-white to light-brown solid, easy to weigh and handle.
  • Storage: store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.