Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2-(2-fluorophenyl)acetic acid | 84145-28-8 | MFCD00042726 | 99.9% | 169.16 g/mol | C8H8FNO2 | 5 G
SDP

Catalog No. 5000299323
Encompass_Preferred

2-Amino-2-(2-fluorophenyl)acetic acid is a fluorinated phenylglycine derivative supplied as a solid for research use. It is provided with high analytical purity and characterized by standard physicochemical identifiers for use in medicinal chemistry and biochemical studies.

  • High purity (99.92%).
  • Molecular weight 169.16 g/mol.
  • Chemical formula C8H8FNO2.
  • CAS number 84145-28-8.
  • Solid appearance suitable for storage and handling in laboratory settings.
  • Intended for research use only; not for human use.
  • Available in multiple package sizes for experimental needs.

Catalog No. 50-002-99323 Supplier Medchemexpress LLC Supplier No. HYW0268945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2-(2-fluorophenyl)acetic acid is a fluorinated phenylglycine derivative supplied as a solid for research use. It is provided with high analytical purity and characterized by standard physicochemical identifiers for use in medicinal chemistry and biochemical studies.

  • High purity (99.92%).
  • Molecular weight 169.16 g/mol.
  • Chemical formula C8H8FNO2.
  • CAS number 84145-28-8.
  • Solid appearance suitable for storage and handling in laboratory settings.
  • Intended for research use only; not for human use.
  • Available in multiple package sizes for experimental needs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.