Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2-(2-methoxyphenyl)acetic acid | 271583-17-6 | MFCD02662408 | ≥97.0% | 181.19 | C9H11NO3 | 25 G
SDP

Catalog No. 5000208808
Encompass_Preferred

2-Amino-2-(2-methoxyphenyl)acetic acid is an amino acid derivative used in research as a building block and intermediate in medicinal chemistry and structure-activity studies. It is supplied as a solid with high purity suitable for laboratory applications.

  • Cas number: 271583-17-6
  • Chemical formula: C9H11NO3
  • Molecular weight: 181.19
  • Purity: ≥97.0%
  • Available sizes: 250 mg to 50 g, including 25 G
  • Grade: For research use only
  • Storage: powder -20°C (3 years) or 4°C (2 years)

Catalog No. 50-002-08808 Supplier Medchemexpress LLC Supplier No. 25GHYW040801
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2-(2-methoxyphenyl)acetic acid is an amino acid derivative used in research as a building block and intermediate in medicinal chemistry and structure-activity studies. It is supplied as a solid with high purity suitable for laboratory applications.

  • Cas number: 271583-17-6
  • Chemical formula: C9H11NO3
  • Molecular weight: 181.19
  • Purity: ≥97.0%
  • Available sizes: 250 mg to 50 g, including 25 G
  • Grade: For research use only
  • Storage: powder -20°C (3 years) or 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.