Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2-(2-methoxyphenyl)acetic acid | 271583-17-6 | MFCD02662408 | ≥97.0% | 181.19 g/mol | C9H11NO3 | 10 G
SDP

Catalog No. 5000209109
Encompass_Preferred

2-Amino-2-(2-methoxyphenyl)acetic acid is an amino-acid-derived research reagent and a glycine derivative used in biochemical and pharmacological studies. It is supplied in multiple laboratory pack sizes and is characterized by its molecular formula and specified purity.

  • Glycine derivative suitable for biochemical research.
  • Molecular formula C9H11NO3; molecular weight 181.19 g/mol.
  • Available in multiple pack sizes up to 50 g.
  • Purity ≥97.0% by supplier specification.
  • CAS number 271583-17-6 for unambiguous identification.

Catalog No. 50-002-09109 Supplier Medchemexpress LLC Supplier No. HYW04080110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2-(2-methoxyphenyl)acetic acid is an amino-acid-derived research reagent and a glycine derivative used in biochemical and pharmacological studies. It is supplied in multiple laboratory pack sizes and is characterized by its molecular formula and specified purity.

  • Glycine derivative suitable for biochemical research.
  • Molecular formula C9H11NO3; molecular weight 181.19 g/mol.
  • Available in multiple pack sizes up to 50 g.
  • Purity ≥97.0% by supplier specification.
  • CAS number 271583-17-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.