Learn More
Medchemexpress LLC 2-amino-2,3-dimethylbutanenitrile | 13893-53-3 | MFCD00661864 | 90.0% | 112.17 | C6H12N2 | 10 G

Supplier: Medchemexpress LLC HYW04080210G
AC-94149 (2-amino-2,3-dimethylbutanenitrile) is an organic nitrile intermediate used in the synthesis of herbicides and agrochemical research. Supplied as a liquid, it is intended for research and compound preparation under controlled storage conditions.
- Molecular formula: C6H12N2.
- Molecular weight: 112.17.
- CAS number: 13893-53-3.
- Purity: 90.0%.
- Physical form: liquid, colorless to light yellow.
- Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
- Packaging sizes: 500 mg, 1 g, 5 g, 10 g, 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.