Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2,3-dimethylbutyronitrile | 13893-53-3 | MFCD00661864 | 90.0% | 112.17 | C6H12N2 | 1 G
SDP

Catalog No. 5000208779
Encompass_Preferred

AC-94149 (2-amino-2,3-dimethylbutyronitrile) is a substituted nitro compound used as an intermediate in the synthesis of several herbicides. The material is supplied at approximately 90.0% purity and is identified by CAS 13893-53-3, molecular formula C6H12N2, and a molecular weight of 112.17 g/mol.

  • used as an intermediate in herbicide synthesis
  • approximate purity 90.0%
  • chemical formula C6H12N2
  • molecular weight 112.17 g/mol
  • available as 1 g package

Catalog No. 50-002-08779 Supplier Medchemexpress LLC Supplier No. HYW0408021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AC-94149 (2-amino-2,3-dimethylbutyronitrile) is a substituted nitro compound used as an intermediate in the synthesis of several herbicides. The material is supplied at approximately 90.0% purity and is identified by CAS 13893-53-3, molecular formula C6H12N2, and a molecular weight of 112.17 g/mol.

  • used as an intermediate in herbicide synthesis
  • approximate purity 90.0%
  • chemical formula C6H12N2
  • molecular weight 112.17 g/mol
  • available as 1 g package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.