Learn More
Medchemexpress LLC 2-amino-2-(3-hydroxyphenyl)acetic acid | 31932-87-3 | MFCD00673854 | 99.6% | 167.16 | C8H9NO3 | 10 MG
SDP
Catalog No. 5000211277
Encompass_Preferred
(Rac)-3-Hydroxyphenylglycine (3HPG) is a racemic amino acid derivative used as a biochemical and pharmacology research reagent. It contains equal parts of the S- and R-enantiomers; the S-enantiomer is a potent, selective metabotropic glutamate receptor 1 (mGluR1) agonist. Supplied as a solid, it is intended for laboratory research applications.
  • Racemic mixture of S- and R-enantiomers.
  • S-enantiomer acts as a selective mGluR1 agonist.
  • Suitable solid form for biochemical and pharmacological assays.
  • High purity (99.58%) as reported by certificate of analysis.
  • Identified by CAS number 31932-87-3 and formula C8H9NO3.
Catalog No. 50-002-11277 Supplier Medchemexpress LLC Supplier No. HY10084110MG
(Rac)-3-Hydroxyphenylglycine (3HPG) is a racemic amino acid derivative used as a biochemical and pharmacology research reagent. It contains equal parts of the S- and R-enantiomers; the S-enantiomer is a potent, selective metabotropic glutamate receptor 1 (mGluR1) agonist. Supplied as a solid, it is intended for laboratory research applications.
  • Racemic mixture of S- and R-enantiomers.
  • S-enantiomer acts as a selective mGluR1 agonist.
  • Suitable solid form for biochemical and pharmacological assays.
  • High purity (99.58%) as reported by certificate of analysis.
  • Identified by CAS number 31932-87-3 and formula C8H9NO3.