Learn More
Medchemexpress LLC 2-Amino-2'-chloro-5-nitrobenzophenone | 2011-66-7 | ≥98.0% | 276.68 | 10 G
SDP
Catalog No. 5000454956
Encompass_Preferred
2-Amino-2'-chloro-5-nitrobenzophenone is described as a precursor in the synthesis of benzodiazepines and a hydrolysis product of Clonazepam.
  • Precursor in the synthesis of benzodiazepines
  • Hydrolysis product of Clonazepam
  • For research use only
Catalog No. 50-004-54956 Supplier Medchemexpress LLC Supplier No. HYW01159610G
2-Amino-2'-chloro-5-nitrobenzophenone is described as a precursor in the synthesis of benzodiazepines and a hydrolysis product of Clonazepam.
  • Precursor in the synthesis of benzodiazepines
  • Hydrolysis product of Clonazepam
  • For research use only