Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-2'-chloro-5-nitrobenzophenone | 2011-66-7 | 98.0% | 276.68 | 25 G
SDP

Catalog No. 5000453296
Encompass_Preferred

2-Amino-2'-chloro-5-nitrobenzophenone serves as a precursor in the synthesis of benzodiazepines. It is also identified as a hydrolysis product of Clonazepam.

  • Purity: 98.0%
  • Molecular weight: 276.68
  • Formula: C13H9ClN2O3
  • CAS no.: 2011-66-7
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-53296 Supplier Medchemexpress LLC Supplier No. 25GHYW011596
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2'-chloro-5-nitrobenzophenone serves as a precursor in the synthesis of benzodiazepines. It is also identified as a hydrolysis product of Clonazepam.

  • Purity: 98.0%
  • Molecular weight: 276.68
  • Formula: C13H9ClN2O3
  • CAS no.: 2011-66-7
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.