Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-2'-deoxyadenosine | 4546-70-7 | MFCD00047240 | 99.6% | 266.26 g/mol | C10H14N6O3 | 1 ML
SDP

Catalog No. 5000303133
Encompass_Preferred

2-Amino-2'-deoxyadenosine is a deoxyribonucleoside analog used as a building block in oligonucleotide synthesis and nucleoside research. It is supplied as a ready-to-use solution in DMSO or as a solid, and is characterized by high purity for laboratory applications.

  • Suitable for oligonucleotide synthesis and nucleoside modification studies.
  • Available as a 10 mM solution in DMSO and in solid pack sizes for scale-up.
  • High purity (99.62%) for research-grade applications.
  • Molecular weight 266.26 g/mol; chemical formula C10H14N6O3.

Catalog No. 50-003-03133 Supplier Medchemexpress LLC Supplier No. HYW01604110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-2'-deoxyadenosine is a deoxyribonucleoside analog used as a building block in oligonucleotide synthesis and nucleoside research. It is supplied as a ready-to-use solution in DMSO or as a solid, and is characterized by high purity for laboratory applications.

  • Suitable for oligonucleotide synthesis and nucleoside modification studies.
  • Available as a 10 mM solution in DMSO and in solid pack sizes for scale-up.
  • High purity (99.62%) for research-grade applications.
  • Molecular weight 266.26 g/mol; chemical formula C10H14N6O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.