Learn More
Medchemexpress LLC 2-Amino-2 -fluoro-5- 100g
SDP
Catalog No. 5000205639
Encompass_Preferred
2-Amino-2 -fluoro-5-nitrobenzophenone is a precursor in the synthesis of benzodiazepines 2-Amino-2 -fluoro-5-nitrobenzophenone is also an acid hydrolysis product of Flunitrazepam
Catalog No. 50-002-05639 Supplier Medchemexpress LLC Supplier No. HYW104076100G
2-Amino-2 -fluoro-5-nitrobenzophenone is a precursor in the synthesis of benzodiazepines 2-Amino-2 -fluoro-5-nitrobenzophenone is also an acid hydrolysis product of Flunitrazepam