Learn More
Medchemexpress LLC 2-Amino-2-methyl-1-propanol | 124-68-5 | 100.0% | 89.14 | 1 ML
SDP
Catalog No. 5000452174
Encompass_Preferred
2-Amino-2-methyl-1-propanol (AMP) acts as a neutralizer, buffer, and surfactant. It is known to inhibit gelatinous product formation and is utilized for carbon dioxide absorption.
  • Neutralizer, buffer, surfactant: versatile chemical with multiple functions.
  • Inhibits gelatinous product formation: prevents the formation of undesirable gelatinous byproducts in non-aqueous CO2 absorbents.
  • Carbon dioxide absorption: used in applications requiring CO2 absorption.
  • Histochemical applications: can be used for histochemical demonstration of alkaline phosphatase activity (at concentrations of 170-350 mM).
  • Safety profile: studies in rats and mice at various doses and exposure routes did not show liver effects or compound-related changes, although some elevated SGPT and OCT serum levels were noted in rats at high oral doses without accompanying gross pathology or histopathology changes.
Catalog No. 50-004-52174 Supplier Medchemexpress LLC Supplier No. HYW01302710MM/1ML
2-Amino-2-methyl-1-propanol (AMP) acts as a neutralizer, buffer, and surfactant. It is known to inhibit gelatinous product formation and is utilized for carbon dioxide absorption.
  • Neutralizer, buffer, surfactant: versatile chemical with multiple functions.
  • Inhibits gelatinous product formation: prevents the formation of undesirable gelatinous byproducts in non-aqueous CO2 absorbents.
  • Carbon dioxide absorption: used in applications requiring CO2 absorption.
  • Histochemical applications: can be used for histochemical demonstration of alkaline phosphatase activity (at concentrations of 170-350 mM).
  • Safety profile: studies in rats and mice at various doses and exposure routes did not show liver effects or compound-related changes, although some elevated SGPT and OCT serum levels were noted in rats at high oral doses without accompanying gross pathology or histopathology changes.