Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Amino-2-methyl-1-propanol

Catalog No. A65182500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65182500ML 500 mL
A651821L 1 L
A6518225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A65182500ML Supplier Sigma Aldrich Supplier No. A65182500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Amino-2-methyl-1-propanol

Specifications

CAS 124-68-5
Melting Point 24°C to 28°C (lit.)
Density 0.934 g/mL (at 25°C (literature))
Boiling Point 165°C (lit.)
Molecular Formula C4H11NO
Refractive Index n20/D 1.4455 (literature)
Linear Formula (CH3)2 C(NH2)CH2OH
MDL Number MFCD00008051
Quantity 500 mL
Synonym beta-Aminoisobutyl alcohol; AMP; AMP 95
Molecular Weight (g/mol) 89.14
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.