Learn More
Medchemexpress LLC 2-Amino-2-methylpropan-1-ol hydrochloride | 3207-12-3 | ≥97.0% | 125.60 | 25 G
SDP
Catalog No. 5000451396
Encompass_Preferred
2-Amino-2-methylpropan-1-ol hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is shipped at room temperature in the continental US and should be stored at room temperature, protected from light. If in solvent, store at -80°C for 1 year or -20°C for 6 months.
  • Biochemical assay reagents
  • Solid appearance
  • White to off-white color
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage at RT, protected from light. In solvent: -80°C for 1 year, -20°C for 6 months
Catalog No. 50-004-51396 Supplier Medchemexpress LLC Supplier No. 25GHYW013027A
2-Amino-2-methylpropan-1-ol hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is shipped at room temperature in the continental US and should be stored at room temperature, protected from light. If in solvent, store at -80°C for 1 year or -20°C for 6 months.
  • Biochemical assay reagents
  • Solid appearance
  • White to off-white color
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage at RT, protected from light. In solvent: -80°C for 1 year, -20°C for 6 months