Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-2-methylpropan-1-ol hydrochloride | 3207-12-3 | ≥97.0% | 125.60 | 500 G
SDP

Catalog No. 5000451999
Encompass_Preferred

2-Amino-2-methylpropan-1-ol hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Purity: ≥97.0%
  • Shipping condition: Room temperature in continental US; may vary elsewhere
  • Storage conditions: RT, protect from light. In solvent -80°C for 1 year, -20°C for 6 months

Catalog No. 50-004-51999 Supplier Medchemexpress LLC Supplier No. HYW013027A500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Amino-2-methylpropan-1-ol hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Purity: ≥97.0%
  • Shipping condition: Room temperature in continental US; may vary elsewhere
  • Storage conditions: RT, protect from light. In solvent -80°C for 1 year, -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.