Learn More
Medchemexpress LLC 2-amino-3-(2'-chloro-5-(phosphonomethyl)-[1,1'-biphenyl]-3-yl)propanoic acid | 174575-17-8 | 98.8% | C16H17ClNO5P | 10MG
SDP
Catalog No. 5000259267
Encompass_Preferred
SDZ 220-581 is a potent, orally active, competitive NMDA receptor antagonist (reported pKi 7.7) provided for research use in neuroscience and pharmacology studies. The compound is supplied with analytical documentation and formulation options to support in vitro and in vivo experiments.
  • Potent NMDA receptor antagonist (pKi 7.7).
  • Orally active pharmacological profile suitable for in vivo studies.
  • Competitive mode of action at NMDA receptors.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High reported purity (≈98.8%).
  • Provided with datasheet, COA, and SDS for quality assurance.
  • Multiple package sizes to accommodate small-scale experiments.
Catalog No. 50-002-59267 Supplier Medchemexpress LLC Supplier No. HY1305910MG
SDZ 220-581 is a potent, orally active, competitive NMDA receptor antagonist (reported pKi 7.7) provided for research use in neuroscience and pharmacology studies. The compound is supplied with analytical documentation and formulation options to support in vitro and in vivo experiments.
  • Potent NMDA receptor antagonist (pKi 7.7).
  • Orally active pharmacological profile suitable for in vivo studies.
  • Competitive mode of action at NMDA receptors.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High reported purity (≈98.8%).
  • Provided with datasheet, COA, and SDS for quality assurance.
  • Multiple package sizes to accommodate small-scale experiments.