Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(2-chlorobenzoyl)-5-ethylthiophene | 50508-60-6 | MFCD04107940 | 98.7% | 265.76 | C13H12ClNOS | 10 G
SDP

Catalog No. 5000301245
Encompass_Preferred

2-amino-3-(2-chlorobenzoyl)-5-ethylthiophene is a chlorobenzoyl-substituted ethylthiophene small molecule supplied for laboratory research and synthetic applications. It is characterized by analytical data and supplied at high purity for use in screening, synthesis, and analytical reference work.

  • CAS registry number 50508-60-6.
  • Molecular formula C13H12ClNOS.
  • Molecular weight 265.76.
  • Physical state: solid.
  • Purity 98.69% (reported).
  • Available in multiple pack sizes suitable for lab use.
  • Useful as a research chemical and synthetic intermediate.

Catalog No. 50-003-01245 Supplier Medchemexpress LLC Supplier No. HYW02493910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-amino-3-(2-chlorobenzoyl)-5-ethylthiophene is a chlorobenzoyl-substituted ethylthiophene small molecule supplied for laboratory research and synthetic applications. It is characterized by analytical data and supplied at high purity for use in screening, synthesis, and analytical reference work.

  • CAS registry number 50508-60-6.
  • Molecular formula C13H12ClNOS.
  • Molecular weight 265.76.
  • Physical state: solid.
  • Purity 98.69% (reported).
  • Available in multiple pack sizes suitable for lab use.
  • Useful as a research chemical and synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.