Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(2-chlorobenzoyl)-5-ethylthiophene | 50508-60-6 | MFCD04107940 | 98.7% | 265.76 g/mol | C13H12ClNOS | 50 G
SDP

Catalog No. 5000299638
Encompass_Preferred

2-Amino-3-(2-chlorobenzoyl)-5-ethylthiophene (CAS 50508-60-6) is a thiophene-derived research chemical described as a benzophenone-like compound. It is offered as a high-purity reagent for use as a synthetic intermediate or analytical reference; confirm lot-specific certificate of analysis for exact purity and handling instructions.

  • High purity (≈98.7%) for analytical and synthetic applications.
  • Molecular formula C13H12ClNOS; molecular weight 265.76 g/mol.
  • Storage at 4°C and protection from light recommended.
  • In-solution stability: -80°C for long-term storage, -20°C for short-term storage.
  • Available in multiple pack sizes to support screening and scale-up.

Catalog No. 50-002-99638 Supplier Medchemexpress LLC Supplier No. HYW02493950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-(2-chlorobenzoyl)-5-ethylthiophene (CAS 50508-60-6) is a thiophene-derived research chemical described as a benzophenone-like compound. It is offered as a high-purity reagent for use as a synthetic intermediate or analytical reference; confirm lot-specific certificate of analysis for exact purity and handling instructions.

  • High purity (≈98.7%) for analytical and synthetic applications.
  • Molecular formula C13H12ClNOS; molecular weight 265.76 g/mol.
  • Storage at 4°C and protection from light recommended.
  • In-solution stability: -80°C for long-term storage, -20°C for short-term storage.
  • Available in multiple pack sizes to support screening and scale-up.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.