Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(3-[(10,12-dicyano-11-(4-(methylcarbamoyl)methoxy)phenyl)carbamoyl]phenyl)propanoic acid | 1941174-13-5 | 99.8% | 1 ML
SDP

Catalog No. 5000231830
Encompass_Preferred

KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter LAT1 (SLC7A5), used in biochemical and cellular research to inhibit leucine uptake and suppress cell growth.

  • Inhibits LAT1-mediated leucine uptake (IC50 = 18 μM).
  • Potentiates the efficacy of bestatin and cisplatin at low concentrations.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Molecular formula C32H25N7O5 and molecular weight 587.58.
  • High purity (99.8%) suitable for research use.

Catalog No. 50-002-31830 Supplier Medchemexpress LLC Supplier No. HY12013910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter LAT1 (SLC7A5), used in biochemical and cellular research to inhibit leucine uptake and suppress cell growth.

  • Inhibits LAT1-mediated leucine uptake (IC50 = 18 μM).
  • Potentiates the efficacy of bestatin and cisplatin at low concentrations.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Molecular formula C32H25N7O5 and molecular weight 587.58.
  • High purity (99.8%) suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.