Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,4,5-trimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A27701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27701G Supplier TCI America Supplier No. A27701G

missing translation for 'validMainframeMsg'

Specifications

CAS 61948-85-4
Melting Point 140°C
Color Yellow
Molecular Formula C10H13NO5
MDL Number MFCD00051630
Quantity 1 g
Synonym 3,4,5-trimethoxyanthranilic acid, benzoic acid,2-amino-3,4,5-trimethoxy, benzoic acid, 2-amino-3,4,5-trimethoxy, maybridge1_001555, acmc-1b683, 2-amino-3,4,5-trimethoxybenzoicacid, 2-amino-3,4,5-trimethoxybenzoic acid
InChI Key JSHSRQCOCMIIPA-UHFFFAOYSA-N
SMILES COC1=C(C(=C(C(=C1)C(=O)O)N)OC)OC
IUPAC Name 2-amino-3,4,5-trimethoxybenzoic acid
Molecular Weight (g/mol) 227.216
PubChem CID 96602
Formula Weight 227.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,4,5-trimethoxybenzoic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.