Learn More
Medchemexpress LLC 2-amino-3-(4-fluorophenyl)propanoic acid | 51-65-0 | MFCD00002600 | 98.8% | 183.18 g·mol⁻¹ | C9H10FNO2 | 10 G
SDP
Catalog No. 5000302296
Encompass_Preferred
2-Amino-3-(4-fluorophenyl)propanoic acid is a phenylalanine derivative used as a research reagent and amino acid analogue for biochemical and medicinal chemistry studies. It is supplied as a white to off-white solid and is accompanied by quality documentation for laboratory use.
  • High purity (98.82%).
  • Solid, white to off-white appearance.
  • Molecular formula C9H10FNO2 and molecular weight 183.18 g·mol⁻¹.
  • Stable as a powder when stored at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Suitable for biochemical research and synthetic applications.
Catalog No. 50-003-02296 Supplier Medchemexpress LLC Supplier No. HYW01634010G
2-Amino-3-(4-fluorophenyl)propanoic acid is a phenylalanine derivative used as a research reagent and amino acid analogue for biochemical and medicinal chemistry studies. It is supplied as a white to off-white solid and is accompanied by quality documentation for laboratory use.
  • High purity (98.82%).
  • Solid, white to off-white appearance.
  • Molecular formula C9H10FNO2 and molecular weight 183.18 g·mol⁻¹.
  • Stable as a powder when stored at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Suitable for biochemical research and synthetic applications.