Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(4-fluorophenyl)propanoic acid | 51-65-0 | MFCD00002600 | 98.8% | 183.18 g/mol | C9H10FNO2 | 25 G
SDP

Catalog No. 5000303092
Encompass_Preferred

2-Amino-3-(4-fluorophenyl)propanoic acid is a research-grade phenylalanine derivative supplied as a white to off-white solid. It is used as a building block in amino acid and peptide synthesis and for biochemical research, provided with characterization and storage guidance for laboratory use.

  • CAS registry number 51-65-0.
  • Molecular formula C9H10FNO2.
  • Molecular weight 183.18 g/mol.
  • Purity 98.82% (typical).
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-003-03092 Supplier Medchemexpress LLC Supplier No. 25GHYW016340
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-(4-fluorophenyl)propanoic acid is a research-grade phenylalanine derivative supplied as a white to off-white solid. It is used as a building block in amino acid and peptide synthesis and for biochemical research, provided with characterization and storage guidance for laboratory use.

  • CAS registry number 51-65-0.
  • Molecular formula C9H10FNO2.
  • Molecular weight 183.18 g/mol.
  • Purity 98.82% (typical).
  • Appearance white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.