Learn More
Medchemexpress LLC 2-amino-3-(4-fluorophenyl)propanoic acid | 51-65-0 | MFCD00002600 | 98.8% | 183.18 g/mol | C9H10FNO2 | 5 G
SDP
Catalog No. 5000302193
Encompass_Preferred
2-Amino-3-(4-fluorophenyl)propanoic acid is a fluorinated phenylalanine derivative provided as a white to off-white solid for laboratory research. It has a molecular weight of 183.18 g/mol and high analytical purity, with storage recommendations to preserve long-term stability.
  • Phenylalanine derivative with a 4-fluoro substitution on the aromatic ring.
  • High purity (98.82%) suitable for research and analytical use.
  • Solid, white to off-white powder for ease of handling and storage.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Supplied for research use only; not intended for human or clinical use.
Catalog No. 50-003-02193 Supplier Medchemexpress LLC Supplier No. HYW0163405G
2-Amino-3-(4-fluorophenyl)propanoic acid is a fluorinated phenylalanine derivative provided as a white to off-white solid for laboratory research. It has a molecular weight of 183.18 g/mol and high analytical purity, with storage recommendations to preserve long-term stability.
  • Phenylalanine derivative with a 4-fluoro substitution on the aromatic ring.
  • High purity (98.82%) suitable for research and analytical use.
  • Solid, white to off-white powder for ease of handling and storage.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Supplied for research use only; not intended for human or clinical use.