Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-bromo-5-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog No. AC437910050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437910050 5 g
AC437910010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437910050 Supplier Thermo Scientific Chemicals Supplier No. 437910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26163-03-1 | Molecular Formula: C5H4BrClN2 | Molecular Weight: 207.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26163-03-1
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.46
MDL Number MFCD00955627
InChI Key UWGGGYYCKDCTGN-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-chloropyridine, 3-bromo-5-chloro-2-pyridinamine, 2-pyridinamine, 3-bromo-5-chloro, 3-bromo-5-chloro-2-pyridylamine, 3-bromo-5-chloropyrid-2-ylamine, 3-bromo-5-chloro-pyridin-2-ylamine, abbypharma ap-10-5406, 3-bromo-5-chloro-pyridin-2-amine, pubchem1141, acmc-1ch3b
PubChem CID 2763971
IUPAC Name 3-bromo-5-chloropyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 82°C to 85°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 207.46
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF INHALED: Remove vic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.