Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. A27991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27991G Supplier TCI America Supplier No. A27991G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Air-sensitive heterocyclic building block for chemical synthesis.

Chemical Identifiers

CAS 21717-95-3
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD04114135
InChI Key WWEINXQNCAWBPD-UHFFFAOYSA-N
Synonym 2-amino-3-fluoropyridine, 3-fluoro-pyridin-2-ylamine, 2-pyridinamine, 3-fluoro, 3-fluoro-2-pyridylamine, 3-fluoro-2-pyridinamine, 3-fluoro-2-aminopyridine, fluoropyridimine, pubchem2976, acmc-209fng, 3-fluoro-pyridin-2ylamine
PubChem CID 2762809
IUPAC Name 3-fluoropyridin-2-amine
SMILES NC1=C(F)C=CC=N1

Specifications

Melting Point 45°C
Boiling Point 110°C
Quantity 1 g
Formula Weight 112.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.