Learn More
Medchemexpress LLC 2-Amino-3-hydroxypyridine | 16867-03-1 | 110.11 | 100 G
SDP
Catalog No. 5000446743
Encompass_Preferred
2-Amino-3-hydroxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can also be used as a chromogenic reagent for the spectrophotometric determination of osmium.
  • Biochemical reagent
  • Used as a biological material or organic compound for life science related research
  • Chromogenic reagent for the spectrophotometric determination of osmium
Catalog No. 50-004-46743 Supplier Medchemexpress LLC Supplier No. HYW007794100G
Add to Cart
Add to Cart
2-Amino-3-hydroxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It can also be used as a chromogenic reagent for the spectrophotometric determination of osmium.
  • Biochemical reagent
  • Used as a biological material or organic compound for life science related research
  • Chromogenic reagent for the spectrophotometric determination of osmium