Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-3-iodopyridine | 104830-06-0 | 99.9% | 220.01 | 10 G
SDP

Catalog No. 5000401444
Encompass_Preferred

2-Amino-3-iodopyridine (3-Iodopyridin-2-amine) can be coupled with formyl derivatives using a copper-catalyzed indole synthesis method, and subsequently, azide analogs can be constructed through copper-mediated conjugated adducts. The nucleophilicity of this compound is lower than that of the corresponding aniline used in indole synthesis. This product is intended for research use only.

  • Can be coupled with formyl derivatives.
  • Azide analogs can be constructed.
  • Solid appearance, off-white to light yellow color.
  • Store at 4°C, protected from light.
  • Soluble in DMSO at 200 mg/mL.

Catalog No. 50-004-01444 Supplier Medchemexpress LLC Supplier No. HY3281310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-iodopyridine (3-Iodopyridin-2-amine) can be coupled with formyl derivatives using a copper-catalyzed indole synthesis method, and subsequently, azide analogs can be constructed through copper-mediated conjugated adducts. The nucleophilicity of this compound is lower than that of the corresponding aniline used in indole synthesis. This product is intended for research use only.

  • Can be coupled with formyl derivatives.
  • Azide analogs can be constructed.
  • Solid appearance, off-white to light yellow color.
  • Store at 4°C, protected from light.
  • Soluble in DMSO at 200 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.