Learn More
Medchemexpress LLC 2-Amino-3-methoxybenzoic acid | 3177-80-8 | 98.0% | C8H9NO3 | 10 G
SDP
Catalog No. 5000434151
Encompass_Preferred
2-Amino-3-methoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. For research use only. We do not sell to patients.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month.
Catalog No. 50-004-34151 Supplier Medchemexpress LLC Supplier No. HY5937710G
2-Amino-3-methoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. For research use only. We do not sell to patients.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month.