Learn More
Medchemexpress LLC 2-Amino-3-methoxybenzoic acid | 3177-80-8 | 98.0% | C8H9NO3 | 5 G
SDP
Catalog No. 5000434207
Encompass_Preferred
2-Amino-3-methoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a solid with a color ranging from off-white to light brown. Shipping is at room temperature in the continental US, with storage recommendations for powder and solvent forms.
  • Can be used as a biological material or organic compound
  • Appears as a solid with off-white to light brown color
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34207 Supplier Medchemexpress LLC Supplier No. HY593775G
2-Amino-3-methoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a solid with a color ranging from off-white to light brown. Shipping is at room temperature in the continental US, with storage recommendations for powder and solvent forms.
  • Can be used as a biological material or organic compound
  • Appears as a solid with off-white to light brown color
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month