Learn More
Medchemexpress LLC 2-amino-3-methyl-3H-imidazo[4,5-f]quinoline | 76180-96-6 | MFCD00210316 | 6980% | 198.22 g/mol | C11H10N4 | 5 MG
SDP
Catalog No. 5000281834
Encompass_Preferred
2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline is a heterocyclic aromatic compound used as a research chemical and reference standard in toxicology and mutagenesis studies. It is reported in literature as a mutagenic heterocyclic amine and is supplied as small, pre-weighed solid quantities for analytical use. Safety documentation including a datasheet and SDS is available from the supplier.
  • Mutagenic heterocyclic amine used in toxicology research.
  • Available in small, pre-weighed solid quantities for analytical assays.
  • High purity ≥98.0% suitable for analytical and reference use.
  • Molecular weight 198.22 g/mol and formula C11H10N4 to support analytical characterization.
  • Safety data and handling instructions provided; follow appropriate precautions.
Catalog No. 50-002-81834 Supplier Medchemexpress LLC Supplier No. HYW2813645MG
2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline is a heterocyclic aromatic compound used as a research chemical and reference standard in toxicology and mutagenesis studies. It is reported in literature as a mutagenic heterocyclic amine and is supplied as small, pre-weighed solid quantities for analytical use. Safety documentation including a datasheet and SDS is available from the supplier.
  • Mutagenic heterocyclic amine used in toxicology research.
  • Available in small, pre-weighed solid quantities for analytical assays.
  • High purity ≥98.0% suitable for analytical and reference use.
  • Molecular weight 198.22 g/mol and formula C11H10N4 to support analytical characterization.
  • Safety data and handling instructions provided; follow appropriate precautions.