Learn More
Medchemexpress LLC 2-amino-3-methyl-3H-imidazo[4,5-f]quinoline | 76180-96-6 | MFCD00210316 | >98.0% | 198.22 | C11H10N4 | 25 MG
SDP
Catalog No. 5000280684
Encompass_Preferred
2-amino-3-methyl-3H-imidazo[4,5-f]quinoline is a mutagenic heterocyclic compound provided for laboratory research and analytical use. It is commonly used as a reference standard in genotoxicity and toxicology studies, and for analytical method development. The substance is identified by CAS 76180-96-6, has formula C11H10N4 and molecular weight 198.22; handle with appropriate safety precautions due to its mutagenic potential.
  • High chemical purity (≥98%) suitable for analytical applications.
  • Available in small milligram pack sizes for targeted experiments.
  • Suitable as a reference standard for genotoxicity and mutagenicity assays.
  • Well characterized by CAS, molecular formula, and molecular weight.
  • Requires handling and disposal under appropriate safety procedures.
Catalog No. 50-002-80684 Supplier Medchemexpress LLC Supplier No. HYW28136425MG
2-amino-3-methyl-3H-imidazo[4,5-f]quinoline is a mutagenic heterocyclic compound provided for laboratory research and analytical use. It is commonly used as a reference standard in genotoxicity and toxicology studies, and for analytical method development. The substance is identified by CAS 76180-96-6, has formula C11H10N4 and molecular weight 198.22; handle with appropriate safety precautions due to its mutagenic potential.
  • High chemical purity (≥98%) suitable for analytical applications.
  • Available in small milligram pack sizes for targeted experiments.
  • Suitable as a reference standard for genotoxicity and mutagenicity assays.
  • Well characterized by CAS, molecular formula, and molecular weight.
  • Requires handling and disposal under appropriate safety procedures.