Learn More
Medchemexpress LLC 2-amino-3-methylimidazo[4,5-f]quinoline | 76180-96-6 | MFCD00210316 | 198.22 g·mol⁻¹ | C11H10N4 | 10 MG
SDP
Catalog No. 5000281451
Encompass_Preferred
2-Amino-3-methylimidazo[4,5-f]quinoline is a heterocyclic aromatic compound used in research on mutagenicity and food-derived carcinogens. It is supplied for laboratory research use only and requires cautious handling due to reported genotoxicity.
  • Used in mutagenicity and toxicology studies.
  • Reported as a food-derived carcinogen relevant to cooked meats and tobacco smoke.
  • Provided as a small-mass solid suitable for analytical and reference use.
  • Identified by CAS 76180-96-6; formula C11H10N4; Mw 198.22 g·mol⁻¹.
  • Suitable for analytical standards and metabolic studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-81451 Supplier Medchemexpress LLC Supplier No. HYW28136410MG
2-Amino-3-methylimidazo[4,5-f]quinoline is a heterocyclic aromatic compound used in research on mutagenicity and food-derived carcinogens. It is supplied for laboratory research use only and requires cautious handling due to reported genotoxicity.
  • Used in mutagenicity and toxicology studies.
  • Reported as a food-derived carcinogen relevant to cooked meats and tobacco smoke.
  • Provided as a small-mass solid suitable for analytical and reference use.
  • Identified by CAS 76180-96-6; formula C11H10N4; Mw 198.22 g·mol⁻¹.
  • Suitable for analytical standards and metabolic studies.
  • For research use only; not for human or clinical use.