Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-methylimidazo[4,5-f]quinoline | 76180-96-6 | MFCD00210316 | >98.0% | 198.22 g/mol | C11H10N4 | 50 MG
SDP

Catalog No. 5000282316
Encompass_Preferred

2-Amino-3-methylimidazo[4,5-f]quinoline is a heterocyclic aromatic amine used as a research reagent in genotoxicity and carcinogenicity studies. It is a known mutagen and is intended for laboratory research use only; follow appropriate safety procedures and consult the safety data sheet for handling instructions.

  • Chemical formula: C11H10N4.
  • Molecular weight: 198.22 g/mol.
  • CAS number: 76180-96-6.
  • Typical purity: >98.0%.
  • Package quantity: 50 MG commonly available.
  • Known hazard: mutagenic compound; use appropriate PPE and containment.

Catalog No. 50-002-82316 Supplier Medchemexpress LLC Supplier No. HYW28136450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-methylimidazo[4,5-f]quinoline is a heterocyclic aromatic amine used as a research reagent in genotoxicity and carcinogenicity studies. It is a known mutagen and is intended for laboratory research use only; follow appropriate safety procedures and consult the safety data sheet for handling instructions.

  • Chemical formula: C11H10N4.
  • Molecular weight: 198.22 g/mol.
  • CAS number: 76180-96-6.
  • Typical purity: >98.0%.
  • Package quantity: 50 MG commonly available.
  • Known hazard: mutagenic compound; use appropriate PPE and containment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.